United Scientific Supplies Specimen Jars, Screw-Cap, Glass
Supplier: GSC International
410-2 410-3 410-4-70-400 410-5
470334-906EA 9.5 CAD
470334-906 470334-908 470334-910 470334-912
United Scientific Supplies Specimen Jars, Screw-Cap, Glass
Jars Specimen Jars
Flint glass specimen jars for easy storage.
  • Autoclavable
All the wide mouth jars are made from soda lime glass and have a PP cap with a PE liner. Since the opening is so large, it is easy to insert and remove materials.
Order Now
SPECIFICATIONS
Environmentally Preferable