Borosil® Reusable Test Tubes without Rim, Foxx Life Sciences
Supplier: Foxx Life Sciences
9820U01 9820U02 9820U03 9820U04 9820U05 9820U06 9820U07 9820U08 9820U09 9820U10 9820U11 9820U12 9820U16 9820U17 9820U18 9820U19
76491-526CS 458.1 CAD
76491-526 76491-528 76491-530 76491-532 76491-534 76491-536 76491-538 76491-540 76491-542 76491-544 76491-546 76491-548 76491-550 76491-552 76491-554 76491-556
Borosil® Reusable Test Tubes without Rim, Foxx Life Sciences
Tubes Test Tubes
Test tubes constructed of borosilicate 3.3 glass, without rim.
  • Excellent chemical resistance
  • Thick walled, mechanically resistant
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Designed without a rim
  • Graduation range: Type I
The large opening can even accommodate large volume funnels, giving easy access to scoops or spatulas.
Amber to protect light-sensitive materials.
Certifications: In compliance with ISO/DIN 4142 standards.