Borosil® Culture Tubes, Clear, PTFE-Lined PP Screw Caps, Foxx Life Sciences
Supplier: Foxx Life Sciences
9900010 9910007
76490-994CS 516.12 CAD
76490-994 76491-014
Borosil® Culture Tubes, Clear, PTFE-Lined PP Screw Caps, Foxx Life Sciences
Tubes Culture Tubes
These Borosil® culture tubes are made of 3.3 borosilicate glass.
  • PTFE lined PP screw caps
  • Round or flat bottom
Certifications: ISO 4142 compliant.