Cole-Parmer® Essentials Pre-Cleaned EPA Narrow Mouth Bottles, Amber Glass, Antylia Scientific
Supplier: Antylia Scientific
99540-32 99540-31 99540-38 99540-33 99540-39 99540-30
76533-950CS 102.79 CAD
76533-950 76533-962 76533-952 76533-964 76533-954 76533-956
Cole-Parmer® Essentials Pre-Cleaned EPA Narrow Mouth Bottles, Amber Glass, Antylia Scientific
Bottles Glass Bottles
Certified clean for worry-free sampling.
  • Includes certificate of conformance to EPA specifications
  • Leakproof PTFE-lined screw caps protect contents
  • Identify samples easier with included analysis labels (unattached)
  • Non-barcode label
  • Autoclavable
These pre-cleaned, narrow-mouth bottles are processed and certified to meet the requirements of the current US EPA 'Specifications and Guidance for Contaminant-free Sample Containers' document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyzes.