Cole-Parmer® Essentials Pre-Cleaned EPA Wide Mouth Septa Jars, Clear Glass, Antylia Scientific
Supplier: Antyila Scientific
99540-20 99540-21
76533-940CS 166.16 CAD
76533-940 76533-942
Cole-Parmer® Essentials Pre-Cleaned EPA Wide Mouth Septa Jars, Clear Glass, Antylia Scientific
Jars General Use Jars
Certified clean for worry-free sampling.
  • Includes certificate of conformance to EPA specifications
  • PTFE-coated silicone septas and caps protect contents
  • Autoclavable
These pre-cleaned, wide mouth jars are processed and certified to meet the requirements of the current US EPA 'Specifications and Guidance for Contaminant-free Sample Containers' document for purgeable volatile organic analysis.