Cole-Parmer® Essentials Pre-Cleaned EPA Wide Mouth Jars, Clear Glass, Antylia Scientific
Supplier: Antyila Scientific
99540-27 99540-40 99540-41 99540-42 99540-43 99540-44
76533-948CS 147.31 CAD
76533-948 76533-966 76533-968 76533-970 76533-972 76533-974
Cole-Parmer® Essentials Pre-Cleaned EPA Wide Mouth Jars, Clear Glass, Antylia Scientific
Jars General Use Jars
Certified clean for worry-free sampling.
  • Includes certificate of conformance to EPA specifications
  • Leakproof PTFE-lined caps protect contents
  • Identify samples easier with included analysis labels (unattached)
  • Autoclavable
These pre-cleaned jars are processed and certified to meet the requirements of the current US EPA 'Specifications and Guidance for Contaminant-free Sample Containers' document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyzes.