Cole-Parmer® Essentials Straight-Sided Round Jars, Clear Glass, Antylia Scientific
Supplier: Antyila Scientific
34800-50 34800-52 34800-54 34800-56 34800-58 34800-60 34800-62
76533-926CS 151.65 CAD
76533-926 76533-928 76533-930 76533-932 76533-934 76533-936 76533-938
Cole-Parmer® Essentials Straight-Sided Round Jars, Clear Glass, Antylia Scientific
Jars General Use Jars
Ideal for general lab use.
  • Leakproof PTFE-lined screw caps protect contents
  • Autoclavable
These straight-sided round bottles allow easy access and transfer of contents.