Cole-Parmer® Essentials Straight-Sided Jars, Amber Glass, Antylia Scientific
Supplier: Antyila Scientific
34700-53 34700-57 34700-65 34700-66
76533-902CS 112.76 CAD
76533-902 76533-904 76533-906 76533-908
Cole-Parmer® Essentials Straight-Sided Jars, Amber Glass, Antylia Scientific
Jars General Use Jars
Don't strain to get samples.
  • Easy access to contents with wide mouth design
  • Store light-sensitive items safely in amber jars
  • Pulp/vinyl lined screw caps protect contents
Manufactured from type III soda-lime glass, these wide mouth jars allow easy access when adding and removing contents. Ideal for general lab use, amber jars are best for light sensitive samples.