Reusable ESD Sheer Labcoats, Short, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
BR3C-42NB-XS BR3C-42NB-S BR3C-42NB-M BR3C-42NB-L BR3C-42NB-XL BR3C-42NB-2XL BR3C-42NB-3XL BR3C-42NB-4XL BR3C-42NB-5XL
76516-264EA 61.13 CAD
76516-264 76516-266 76516-268 76516-270 76516-272 76516-274 76516-276 76516-278 76516-280
Reusable ESD Sheer Labcoats, Short, ESD Knit Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 91% polyester/9% carbon.
  • Lapel collar
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.