Reusable Maxima ESD Pull-Over Cleanroom Hoods, Uniform Technology
Supplier: Uniform Technology
CHPO-68WH-XS CHPO-68WH-S CHPO-68WH-M CHPO-68WH-L CHPO-68WH-XL CHPO-68WH-2XL CHPO-68WH-3XL
76515-916EA 25.55 CAD
76515-916 76515-918 76515-920 76515-922 76515-924 76515-926 76515-928
Reusable Maxima ESD Pull-Over Cleanroom Hoods, Uniform Technology
Workwear
Cleanroom hoods are manufactured from 99% polyester/1% carbon fabric.
  • Pull-over, open face design
  • Vertical and horizontal snaps on back
  • Snaps at temples
  • Split skirt
Suitable for ISO Class 4 or higher cleanrooms.