Reusable Altessa Grid Cleanroom Boots with Hypalon Sole, Uniform Technology
Supplier: Uniform Technology
CB3-74WH-XS CB3-74WH-S CB3-74WH-M CB3-74WH-L CB3-74WH-XL CB3-74WH-2XL CB3-74WH-3XL
76515-854PR 55.77 CAD
76515-854 76515-856 76515-858 76515-860 76515-862 76515-864 76515-866
Reusable Altessa Grid Cleanroom Boots with Hypalon Sole, Uniform Technology
Workwear
Cleanroom boots are manufactured from 99% polyester/1% carbon fabric.
  • Color-coded by size webbing strap over toe and ankle
  • Side release buckle
  • Hypalon sole
Suitable for ISO Class 5 or higher cleanrooms.