Reusable Altessa Grid Cleanroom Frocks with Zipper Closure, Uniform Technology
Supplier: Uniform Technology
CFRZC-74WH-XS CFRZC-74WH-S CFRZC-74WH-M CFRZC-74WH-L CFRZC-74WH-XL CFRZC-74WH2XL CFRZC-74WH3XL CFRZC-74WH4XL CFRZC-74WH5XL CFRZC-74NV-XS CFRZC-74NV-S CFRZC-74NV-M CFRZC-74NV-L CFRZC-74NV-XL CFRZC-74NV2XL CFRZC-74NV3XL CFRZC-74NV4XL CFRZC-74NV5XL CFRZC-74LB-XS CFRZC-74LB-S CFRZC-74LB-M CFRZC-74LB-L CFRZC-74LB-XL CFRZC-74LB2XL CFRZC-74LB3XL CFRZC-74LB4XL CFRZC-74LB5XL
76515-684EA 61.54 CAD
76515-684 76515-686 76515-688 76515-690 76515-692 76515-694 76515-696 76515-698 76515-700 76515-702 76515-704 76515-706 76515-708 76515-710 76515-712 76515-714 76515-716 76515-718 76515-720 76515-722 76515-724 76515-726 76515-728 76515-730 76515-732 76515-734 76515-736
Reusable Altessa Grid Cleanroom Frocks with Zipper Closure, Uniform Technology
Workwear
Cleanroom frocks are manufactured from 99% polyester/1% carbon fabric.
  • Military collar
  • Raglan sleeves
  • ESD knit wrists
  • Coil zipper with top snaps
Suitable for ISO Class 5 or higher cleanrooms.
Order Now
SPECIFICATIONS
Model CFRZC-74
Environmentally Preferable
Garment application Cleanroom
Risk area
Gender
Closure type Zip closure
Fabric 99% polyester/1% carbon