Cole-Parmer® Essentials Economy Amber Wide-Mouth Plastic Bottle, HDPE, Antylia Scientific
Supplier: Antylia Scientific
06036-05 06036-11 06036-09 06036-15 06036-01 06036-13 06036-07 06036-21 06036-19 06036-23 06036-03 06036-17
76534-172PK 432.43 CAD
76534-172 76534-162 76534-170 76534-160 76534-180 76534-158 76534-178 76534-168 76534-176 76534-166 76534-174 76534-164
Cole-Parmer® Essentials Economy Amber Wide-Mouth Plastic Bottle, HDPE, Antylia Scientific
Bottles Plastic Bottles
Keep light-sensitive samples safe.
  • Reduce UV light transmission
  • Meets USP Class VI requirements
  • Will not leak with PP screw caps
Protect light-sensitive materials during shipping and storage. Amber bottles reduce UV light transmission and meet USP requirements for storage of light-sensitive materials. Suitable for packing, storage, and shipping liquids. All bottles have leakproof polypropylene screw caps. Bottles are not autoclavable.