Cole-Parmer® Essentials Economy Narrow-Mouth Plastic Bottles HDPE, Antylia Scientific
Supplier: Antyila Scientific
06035-07 06035-09 06035-11 06035-13 06035-15 06035-17 06035-19 06035-21 06035-23 06035-25 06035-27 06035-29
76534-088PK 18.65 CAD
76534-088 76534-090 76534-094 76534-096 76534-098 76534-100 76534-102 76534-106 76534-108 76534-110 76534-112 76534-114
Cole-Parmer® Essentials Economy Narrow-Mouth Plastic Bottles HDPE, Antylia Scientific
Bottles Plastic Bottles
Chemical-resistant bottles stand up to most corrosives.
  • Withstands freezer storage down to −100 °C
  • Will not leak with PP screw caps
  • Meets USP Class VI requirements
These all-purpose bottles are popularly used for packaging, shipping, and storage because of their reliable durability. Rigid, translucent, and chemical-resistant, they can be used with most corrosives. All have leakproof polypropylene screw caps. Bottles are not autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable