Cole-Parmer® Essentials Pre-Cleaned Wide-Mouth Plastic Bottle, HDPE, Antylia Scientific
Supplier: Antyila Scientific
62266-10 62266-12 62266-14 62266-16
76534-346CS 373.68 CAD
76534-346 76534-348 76534-350 76534-352
Cole-Parmer® Essentials Pre-Cleaned Wide-Mouth Plastic Bottle, HDPE, Antylia Scientific
Bottles Plastic Bottles
Leakproof design keeps samples safe.
  • Pre-cleaned according to EPA standards
  • Barcoded label type
These wide-mouth, high-density polyethylene (HDPE) bottles are leak-proof and break-resistant. They are excellent for soil, sludge, and sediment samples. Bottles are pre-cleaned according to Environmental Protection Agency (EPA) standards and certified to meet the EPA’s Performance Based Specifications for metals analysis and water quality parameters. Bottles are not autoclavable.