Cole-Parmer® Essentials Amber Narrow-Mouth Plastic Bottles, PP, Antylia Scientific
Supplier: Antyila Scientific
06027-10 06027-20
76534-026EA 52.88 CAD
76534-026 76534-028
Cole-Parmer® Essentials Amber Narrow-Mouth Plastic Bottles, PP, Antylia Scientific
Bottles Plastic Bottles
Store chemical samples safely.
  • Leakproof polypropylene caps
  • Built-in shoulder loop for easier handling
  • Autoclavable
Use these sturdy bottles whenever excellent chemical resistance is desired for most acids, bases, and alcohols. Protects light-sensitve liquids by reducing UV light transmissions. Ideal for storing chemicals and standards, as well as for collecting and storing distilled water. Larger bottles feature built-in shoulder loop.