Cole-Parmer® Deep-Well Microplates and Sealing Mats for Homogenizers
Supplier: SPEX
2220 2220-100 2200 2200-100 2210 2210-100 2230 2230-100 2205-50
76181-824EA 48.05 CAD
76181-824 76181-826 76182-264 76182-266 76182-280 76182-282 76182-288 76182-290 76533-132
Cole-Parmer® Deep-Well Microplates and Sealing Mats for Homogenizers
Microplates Deep Well Microplates
Durable polypropylene microplates resist perforation by grinding media.
  • Alphanumeric marks for well identification
  • Use microplates with sealing mats to prevent spills and well-to-well contamination
Ideal for use with Cole-Parmer® homogenizers, these deep-well plates serve many application purposes, such as tissue homogenization and cell lysis.
Load each well with the sample, solution (if desired), and grinding media then seal with a sealing mat to prevent spills and cross contamination.
The sealing mats are constructed of rugged silicone rubber and designed to be sterilized and reused.