2010 Geno/Grinder® Homogenizer Adjustable Clamp, SPEX SamplePrep
Supplier: SPEX SamplePrep
2011
76533-110EA 4485 CAD
76533-110
2010 Geno/Grinder® Homogenizer Adjustable Clamp, SPEX SamplePrep
Homogenizers Benchtop Homogenizers
Adjusts to fit various sizes of titer plates and vials.
  • Accommodates up to six deep-well titer plates or up to sixteen 50 ml tubes
  • Stack titer plates in the clamp increasing throughput
  • Release mechanism makes it easy to slide clamp up and down the shaft
  • Adjustable Clamp Assembly for new model 2010 Geno/Grinder
  • Clamp provides 1.25" (3.2 cm) vertical movement
This front loading clamp is included as standard in all new 2010 Geno/Grinders but can be purchased separately as a retro-fit.