Vikan® High Temperature Scrapers, Remco
Supplier: Remco Products
69632 69633 69634 69635 69636
76530-234EA 17.47 CAD
76530-234 76530-236 76530-238 76530-240 76530-242
Vikan® High Temperature Scrapers, Remco
Scrapers
These high temperature large hand scrapers are great for scraping hot cooking surfaces.
  • For use in temperatures up to 410 °F
  • Nylon/glass fiber construction
  • Seamless design
  • Length: 9.38"
The scraper has no seams or cracks, which helps prevent bacterial growth.
Order Now
SPECIFICATIONS
Environmentally Preferable