Vikan® Pipe Brush Heads, Medium Bristles, Remco
Supplier: Remco Products
53801032 53801033 53801034 53801035 53801036 53801039 5380772 5380773 5380774 5380775 5380776 5380779 5380902 5380903 5380904 5380905 5380906 5380909
76530-092EA 46.7 CAD
76530-092 76530-094 76530-096 76530-098 76530-100 76530-102 76530-124 76530-126 76530-128 76530-130 76530-132 76530-134 76530-136 76530-138 76530-140 76530-142 76530-144 76530-146
Vikan® Pipe Brush Heads, Medium Bristles, Remco
Brushes
This tube/pipe brush head has bristles along the sides as well as in the front. It is used in areas where a wider tube cleaning action is required or where the equipment has an end point that cannot be reached with a hand brush.
  • Polyester bristles, polypropylene block
  • Euro (hygienic) thread type
  • Available in 3.0", 3.5" and 4.0" Ø
Works well as a drain brush, particularly in black. Requires a European thread handle.