Vikan® UST Bench Brushes, Medium Bristles, Remco
Supplier: Remco Products
45851 45852 45853 45854 45855 45856 45857 45858
76530-014EA 46.06 CAD
76530-014 76530-016 76530-018 76530-020 76530-022 76530-024 76530-026 76530-028
Vikan® UST Bench Brushes, Medium Bristles, Remco
Brushes
This UST bench brush with medium strength bristles is ideal for sweeping damp particles from conveyor belts, food preparation surfaces, tables, and equipment in high risk areas.
  • Color-coded to support HACCP principles
  • Polypropylene block, polyester bristles
The placement of the bristles enables efficient sweeping of damp and wet particles.
Overall dimensions: 13L×1.5W×3.9"H