Vikan® Baker's Brushes, Soft Bristles, Remco
Supplier: Remco Products
45822 45823 45824 45825 45826
76530-004EA 31.81 CAD
76530-004 76530-006 76530-008 76530-010 76530-012
Vikan® Baker's Brushes, Soft Bristles, Remco
Brushes
This long, narrow,fully color-coded hand brush is perfect for dusting large surface areas.
  • Color-coded to support HACCP principles
  • Polypropylene block, polyester bristles
Soft bristles are designed for brushing dry, fine particles like dust, flour, and powdered sugar.
Overall dimensions: 11L×1.2W×3.7"H