Vikan® UST Long Handle Scrubbing Brushes, Stiff Bristles, Remco
Supplier: Remco Products
41961 41962 41963 41964 41965 41966 41967 41968
76528-942EA 53.7 CAD
76528-942 76528-944 76528-946 76528-948 76528-950 76528-952 76528-954 76528-956
Vikan® UST Long Handle Scrubbing Brushes, Stiff Bristles, Remco
Brushes
This UST long handled hand brush with stiff bristles is effective when cleaning machines, buckets, containers, and tubs in high risk areas.
  • Color-coded to support HACCP principles
  • Polypropylene block, polyester bristles
The angled and circular bristle arrangement enables efficient scrubbing and washing process.
Overall dimensions: 15.5L×2.875W×2.75"H