Vikan® Nylon Scrapers with Threaded Handle, Remco
Supplier: Remco Products
40122 40123 40124 40125 40126
76528-894EA 24.85 CAD
76528-894 76528-896 76528-898 76528-900 76528-902
Vikan® Nylon Scrapers with Threaded Handle, Remco
Scrapers
Suitable for loosening stubborn dirt like pastry, chocolate, or burnt food on smooth surfaces.
  • Nylonl blade, polypropylene grip
  • Euro (hygienic) thread type
The nylon blade is a good alternative to a stainless steel, and can be used on sensitive surfaces to avoid scratches on equipment and sensitive conveyor belts. The hard blade can withstand hot surfaces when used in two minute intervals. Can be used with any Vikan handle.
Order Now
SPECIFICATIONS
Environmentally Preferable