Vikan® 16" UST Push Broom Head, Medium Bristles, Remco
Supplier: Remco Products
31731 31732 31733 31734 31735 31736 31737 31738
76528-838EA 95.92 CAD
76528-838 76528-840 76528-842 76528-844 76528-846 76528-848 76528-850 76528-852
Vikan® 16" UST Push Broom Head, Medium Bristles, Remco
Brooms
The 16" Ultra Safe Technology (UST) medium push broom is effective for sweeping in dry or wet high risk areas.
  • Polypropylene block, polyester bristles
  • Euro (hygienic) thread type
The angled arrangement of the Bristle Security Units ensures efficient and effective cleaning, while the lack of resin, glue, or staples means bristle units are easy to clean, FDA-compliant, and more secure.
Broom head dimensions: 15.25L×1.875W×4.7"H