Vikan® 51" Aluminum Handle, Remco
Supplier: Remco
Vikan®
293566 293588 293577
76528-748EA 49.6 CAD
76528-748 76528-746 76528-750
Vikan® 51" Aluminum Handle, Remco
Brooms
Standard broom handle can be used with all brooms, squeegees and scarpers.
  • Aluminum construction with color-coded polypropylene grips
  • Euro (hygienic) thread type
Also suitable for use as a broom handle. It is recommended the handle with attached item should reach the chin of user to prevent back injuries.