Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Supplier: Remco Products
10121 10122 10123 10124 10125 10126 101266 10127 101277 10128 101288 10129
76528-624EA 20 CAD
76528-624 76528-626 76528-628 76528-630 76528-632 76528-634 76528-636 76528-638 76528-640 76528-642 76528-644 76528-646
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Wall Hooks
Single hook module is designed for storing cleaning tools that have a hanging hole.
  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction
The hook system is easy to disassemble for cleaning or replacement.