FormaPure XL Reagent Kit, Beckman Coulter

Supplier: BECKMAN COULTER CANADA LP CA

C35991 C35992
CA76409-741EA 1015.35 CAD
CA76409-741 CA76409-743
FormaPure XL Reagent Kit, Beckman Coulter
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

RNA and DNA extraction from same lysate of FFPE tissues. The FormaPure XL Total has been demonstrated on four tissue types.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Target molecule
Refine target molecule
Sample type FFPE tissues
Method / Format

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR