GenFind V3 Reagent Kits, Beckman Coulter
Supplier: BECKMAN COULTER CANADA LP CA
C34880 C34881
CA76409-759EA 331.98 CAD
CA76409-759 CA76409-761
GenFind V3 Reagent Kits, Beckman Coulter
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
This prep system kit isolates and purifies genomic DNA (gDNA) from whole blood. The kit is uses Solid Phase Reversible Immobilization (SPRI) paramagnetic bead-based technology to effectively produce a high recovery of high quality DNA for downstream applications such as PCR.
  • Automated available