DNAdvance Kits, Beckman Coulter
Supplier: BECKMAN COULTER CANADA LP CA
A48705 A48706
CABKA48705EA 2050.26 CAD
CABKA48705 CABKA48706
DNAdvance Kits, Beckman Coulter
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Isolation from mammalian tissue. Built on SPRI paramagnetic bead-based technology, the DNAdvance Kit is a high throughput genomic DNA (gDNA) isolation reagent kit enabling the purification of high quality DNA from mammalian tissue samples.
  • Automated available