RNAdvance Blood Kits, Beckman Coulter
Supplier: BECKMAN COULTER CANADA LP CA
A35604 A35603
CABKA35604EA 4957.45 CAD
CABKA35604 CA76522-026
RNAdvance Blood Kits, Beckman Coulter
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Built on SPRI (Solid Phase Reversible Immobilization) paramagnetic bead-based technology, the RNAdvance Blood Kit is a ribonucleic acid (RNA) isolation process that enables you to purify high quality RNA from blood you’ve collected using PAXgene tubes.
  • Automated available