Abdos® Low Retention Racked Filter (Barrier) Micro Tips, Graduated, Gamma Sterilized, Foxx Life Sciences
Supplier: Foxx Life Sciences
P11065 P11067 P11075
76516-490CS 259.71 CAD
76516-490 76516-492 76516-494 76516-496 76516-498 76516-500 76516-502 76516-504 76516-508
Abdos® Low Retention Racked Filter (Barrier) Micro Tips, Graduated, Gamma Sterilized, Foxx Life Sciences
Pipette Tips
Abdos® low retention filter tips are made of hydrophobic polypropylene (PP) conforming to US FDA 21CFR. The filters are 100% pure porous (PE) which allows recovery of wetted samples while the inert, hydrophobic filter does not trap or block liquids.
  • Natural color
  • Sterilization by gamma radiation
  • Temperature range: 0 to 135 °C
  • Three years shelf life
  • Animal-free
  • Universal fit
The filter prevents liquids and aerosols from passing through the filter and elimates contamination of the pipette. Polypropylene racks are very robust, sturdy with thick wallMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable