Medicor® Infant Foot For Heel Prick Screening Test
Supplier: ELEVATE HEALTHCARE, INC. SE
Medicor®
M-NNPES-001-B M-NNPES-001-N M-NNPES-001-M
470337-035EA 405.2 CAD
470337-035 470337-036 470337-037
Medicor® Infant Foot For Heel Prick Screening Test
Models Human Biology Models
The only heel prick trainer available.
  • Contains internal reservoir
  • Available in three skin tones
Performing infant blood screenings is crucial to detect rare but serious disorders that may affect a baby's health. Students can learn and practice the appropriate technique using this infant foot that features lifelike blood sampling. The model realistically simulates the softness and structure of a baby's heel while the internal reservoir releases a small trickle of blood with the application of light pressure.