Medicor® Injectable Abdomen
Supplier: ELEVATE HEALTHCARE, INC. SE
I-A-001-S-PR-B I-A-001-S-PR-M I-A-001-S-PR-N
470336-964EA 431.7 CAD
470336-964 470336-966 470336-968 470336-970 470336-972 470336-974
Medicor® Injectable Abdomen
Models Human Biology Models
Practice subcutaneous injection.
  • Available in three skin tones
  • Available in a belted version
Made entirely of proprietary MedicSkin, injectable abdominal models provide a lifelike experience to practice administering subcutaneous injections. Students get a realistic haptic experience squeezing the tissue and placing the injection. The interior of the model contains a sponge, making it ideal to practice injecting fluid into the model without deterioration or molding. Choose from the base model or the belted model with protection plate. The protection plate provides a barrier to avoid accidental punctures while the belt allows learners to securely attach to manikins or standardized patients.