Medicor® Peripheral Intravenous (IV) Catheterization Arm Model
Supplier: ELEVATE HEALTHCARE, INC. SE
IV-BT-001-TC-B IV-BT-001-TC-M IV-BT-001-TC-N
470336-962EA 2123.55 CAD
470336-962 470337-041 470337-042
Medicor® Peripheral Intravenous (IV) Catheterization Arm Model
Models Human Biology Models
Perform IV and IM on a single model.
  • Available in three skin tones
  • Mold resistant
  • Self healing skin to 22G
A best-selling product, this IV arm features four veins to practice performing peripheral IV insertions. Each arm comes with two 1000 ml bags to use for simulating an IV drip and blood flow through the arm. After inserting the catheter correctly, students experience a realistic flashback for placement confirmation.
Caution: Some components of this product may contain latex.