HIV-1 Trimeric GP140 Protein-JRFL Strain
Supplier: ReVacc Scientific
G87X55-100UG
G87X55-500UG
76519-270EA
1199
CAD
76519-270
76519-272
HIV-1 Trimeric GP140 Protein-JRFL Strain
Proteins and Peptides
A recombinant trimeric form of the HIV-1 glycoprotein (GP protein) from JRFL strain (GenBank: AAB05604) was produced by human embryonic kidney HEK293F cells, followed by Lectin purification.
- Protein function: Bind to its receptor
This product contains ectodomain of GP protein with furin cleavage site knocked-down, T4 foldon trimerization domain, and C-terminal 6 hexa-histidine tag.
This product has been evaluated by native blue gel and ELISAs.