HIV-1 Trimeric GP140 Protein-YU2 Strain

Supplier: ReVacc Scientific

W44H24-100UG W44H24-500UG
76519-266EA 1199 CAD
76519-266 76519-268
HIV-1 Trimeric GP140 Protein-YU2 Strain
Proteins and Peptides

A recombinant trimeric form of the HIV-1 glycoprotein (GP protein) from YU2 strain (GenBank: QHJ92204) was produced by human embryonic kidney HEK293F cells, followed by Lectin purification.


  • Protein function: Bind to its receptor


This product contains ectodomain of GP protein with furin cleavage site knocked-down, T4 foldon trimerization domain, and C-terminal 6 hexa-histidine tag.


This product has been evaluated by native blue gel and ELISAs.

Order Now

SPECIFICATIONS

Protein/peptide name HIV-1 trimeric GP140 protein-YU2 strain
Environmentally Preferable
Protein synonyms HIV-1 trimeric GP140 PROTEIN-YU2 strain (His Tag)
Protein/peptide type Recombinant
Species Virus
Source Mammalian cells
Conjugation Tag conjugated
Gene ID QHJ92204
Protease-free No
Animal-free Yes
Nuclease-free No
Carrier-free Yes
Endotoxin-free No
Endotoxin level Not detected
Purity 60 - 85%
Concentration 0.2 mg/ml
Formulation Frozen liquid. In PBS pH 7.5
Molecular weight ~76 kDa
Storage conditions –80 °C
Shipping temperature 70
Tested applications ELISA, NativeBlue

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR