SP Bel-Art Buchner Table-Top Funnels, 610 mm (24"), Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H14629-0000 H14622-0000 H14625-3524 H14627-1610 H91462-4361 H91462-4610 H91463-1610
13197-330EA 2969.14 CAD
13197-330 30308-094 47750-790 47750-800 47750-808 47750-816 74950-370 74950-374 74950-382
SP Bel-Art Buchner Table-Top Funnels, 610 mm (24"), Bel-Art Products, a part of SP
Funnels Filtration Funnels Büchner Funnels
Filter large volumes quickly with a polyethylene plastic Table-Top Buchner Funnel using your choice of Fritware® or Perforated filter plate.
  • Removable hose barbed connector accepts 12.7 mm (½") I.D. tube and threads into ¾" NPT bung on the funnel
  • Overall height: 330 mm (13")
  • Rim to plate: 254 mm (10")
Fritware® Porous Filter Plates supported by a multiple ring grid, are made of 6.4 mm (¼") thick HDPE, with a non-porous ring at the periphery of the plate which seals filter paper when used. Fritware® Porous Filter Plates (fixed or removable) areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable