SP Bel-Art Buchner Table-Top Funnels, 914 mm (36"), Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H14630-0000 H14623-0000 H14625-3536 H14626-3536 H14627-1914 H91462-4091 H91462-4914 H91463-1914
13197-332EA 6578.58 CAD
13197-332 16126-608 47750-794 47750-802 47750-810 47750-818 74950-364 74950-376 74950-384
SP Bel-Art Buchner Table-Top Funnels, 914 mm (36"), Bel-Art Products, a part of SP
Funnels Filtration Funnels Büchner Funnels
Filter large volumes quickly with a polyethylene plastic Table-Top Buchner Funnel using your choice of Fritware® or Perforated filter plate.
  • Removable hose barbed connector accepts 12.7 mm (½") I.D. tube and threads into ¾" NPT bung on the funnel
  • Overall height: 350 mm (13.75")
  • Rim to plate: 318 mm (12.5")
Fritware® Porous Filter Plates supported by a multiple ring grid, are made of 6.4 mm (¼") thick HDPE, with a non-porous ring at the periphery of the plate which seals filter paper when used. Fritware® Porous Filter Plates (fixed or removable) areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable