SynaptoGreen™ C18 (Equivalent to FM®3-25) membrane stain
Supplier: Biotium
70048 70049
89139-454EA 781.61 CAD
89139-454 89139-456
SynaptoGreen™ C18 (Equivalent to FM®3-25) membrane stain
SynaptoGreen™ C18 (Equivalent to FM®3-25)
SynaptoGreen™ C18 (equivalent to FM®3-25) is a membrane stain with a chemical structure similar to other nerve terminal dyes, except that it has the longest lipophilic tail (C18). Because of its large size, it is used as a control for ion channel permeability of nerve terminal dyes.
  • Red solid soluble in DMSO or DMF
  • Ex/Em: 510/625 nm (in MeOH); 480/598 nm (in membranes)
SynaptoGreen™ C18 (equivalent to FM®3-25) is a membrane stain with a chemical structure similar to other nerve terminal dyes, except that it has the longest lipophilic tail (C18). Because of its large size, it is used as a control for ion channel permeability of nerve terminal dyes.
FM is a trademark of Molecular Probes, Inc
Caution: Store desiccated at 4°C and protect from light, especially in solution
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c423e48df6d9395f59f5bf333934bd19b6b89743&vid=84b1b0df3d1bd74898dec48c5c993af3cc597599&prid=web&clid=SAPDAM