VWR® Glassware Washer Spindle Racks
Supplier: VWR International
10002-946 10002-948 10002-950
10002-946EA 9041.31 CAD
10002-946 10002-948 10002-950
VWR® Glassware Washer Spindle Racks
Glassware Washers
Useful for cleaning inside narrow-neck glassware, these spindle racks are designed for use with undercounter, freestanding and Vertical Stacking VWR® Glassware Washer models.
  • Recommended for graduated cylinders, volumetric flasks, round bottom flasks, Erlenmeyer flasks and other narrow-neck glassware
  • Multiple configurations available to suit a variety of glassware
See details in table below for specific use information. For further assistance, view the product literature or contact your Avantor representative
Note: Spindle racks fit only in unitsMore Product Information