Cole-Parmer Stuart Hybrigene and Hybridizer Incubators, Antylia Scientific
Supplier: Antyila Scientific
39455-01 39455-07 39455-08
76531-222EA 12257.06 CAD
76531-222 76531-224 76531-226
Cole-Parmer Stuart Hybrigene and Hybridizer Incubators, Antylia Scientific
Ovens Hybridisation Ovens
Use and recover volumes as low as 5 ml, ensuring preservation of precious samples.
  • Radioactive - safe protective casing and non-drip tube design minimize risk to user
  • Temperature resolution: 0.1 °C
With excellent stability of ±0.1 °C within the tubes, these Hybrigene and Hybridizer incubators are great for nucleic hybridization applications such as Northern, Southern, and Western blots. Each incubator features two shakers - rotisserie shaker with rotational speed from 0 to 20 rpm and platform shaker with rocking speed up to 60 oscillation/minute. AMore Product Information