Kimtech™ Vista™ Nitrile Examination Gloves, Kimberly-Clark
Supplier: Microflex
62825 62826 62827 62828 62829
76520-546CS 700.72 CAD
76520-546 76520-548 76520-550 76520-552 76520-554
Kimtech™ Vista™ Nitrile Examination Gloves, Kimberly-Clark
Gloves
Choose Vista™ nitrile examination gloves by Kimberly-Clark Professional™ to reduce laboratory waste while providing excellent glove dexterity, strength, and protection. 
  • Ambidextrous, 9.5" nitrile exam gloves
  • Beaded cuffs prevent roll-down
  • Strong grip (wet or dry), with textured fingertips for dexterity
  • AQL 1.5 (exceeds the ASTM 2.5 standard for critical defects or pinholes)
  • Food Handling Certified in the United States
These nitrile gloves have a strong wet and dry grip, which reduces the potential for slips. Plus, the thin mil gloves texturedMore Product Information