Kimtech™ Element™ Nitrile Examination Gloves, Kimberly-Clark
Supplier: Microflex
62870 62871 62872 62873 62874
76520-536CS 713.54 CAD
76520-536 76520-538 76520-540 76520-542 76520-544
Kimtech™ Element™ Nitrile Examination Gloves, Kimberly-Clark
Gloves
These reliable, easy-to-wear gloves are more economical than standard nitrile gloves and are suited for a wide variety of tasks.
  • Ambidextrous and powder-free
  • Beaded cuffs
  • 3.2 mil thickness
  • Superior quality, AQL 1.5 (exceeds the ASTM D 5151 standard for critical defects or pinholes)
  • Not made with natural rubber latex
  • Waste less with the one-at-a-time dispenser box
  • 250% more gloves per case (versus standard nitrile gloves)
The search for a comfortable, protective exam glove with exceptional value is over, thanks to Kimtech™ Element™ nitrile examinationMore Product Information