Kimtech™ G3 EvT Prime™ Nitrile Gloves, Kimberly-Clark
Supplier: Microflex
62005 62006 62007 62008 62009
76501-046CS 720.04 CAD
76501-046 76501-048 76501-050 76501-052 76501-054
Kimtech™ G3 EvT Prime™ Nitrile Gloves, Kimberly-Clark
Gloves
White nitrile cleanroom gloves.
  • Non sterile, ambidextrous glove with textured fingertips
  • 4.72 mil / 0.12 mm thickness
  • Beaded cuff
  • Amide-free, powder-free
These gloves are double chlorinated and double bagged. These gloves are not made with silicone.
Recyclable via the RightCycle™ program.