KleenGuard™ G10 Comfort Plus™ Nitrile Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
54185 54186 54187 54188 54189
CA76501-066BX 48.64 CAD
CA76501-066 CA76501-068 CA76501-070 CA76501-072 CA76501-074
KleenGuard™ G10 Comfort Plus™ Nitrile Gloves, Kimberly-Clark
Gloves
When searching for a glove that is designed for superior strength and tactile sensitivity, KleenGuard™ G10 Comfort Plus™ nitrile gloves are an excellent choice.
  • Light blue nitrile gloves
  • Powder-free, ambidextrous, and touchscreen compatible
  • Comfortable, protective, and user-friendly
  • Advanced formulation delivers superior strength and tactile sensitivity
KleenGuard™ Personal Protective Equipment (PPE) enables rather than inhibits, allowing for maximum productivity and safety for you and your employees. These 9" ambidextrous gloves feature a beaded cuff (for easy donning), textured fingertips (for dexterity and grip), and 4 mil thickness for comfort and ease of movement. They are offered in a variety of sizes, and are not made with natural rubber latex.
These food grade gloves meet U.S. FDA regulations for food contact applications, food handling and prep, and they are also good for general manufacturing, food service, electrical assembly, electrical contracting, painting, aluminum die casting, and automotive uses. Choose these gloves for the protection of nitrile at a great everyday price.
G10 Comfort Plus™ blue gloves can be recycled via the Kimberly Clark Professional RightCycle™ program.