VWR® Freeze Spray

Supplier: VWR
VWR®
78907-09
76528-072EA 76.69 CAD
76528-072
VWR® Freeze Spray
Freezing Sprays
Instantly cool electrical components to –65 °F.

  • Dries fast and leaves no residue
  • Quickly deliver freezing power to hard-to-reach places
  • Ultra-low global warming potential (GWP)
  • Nonflammable and ozone safe

VWR® Freeze spray is a nonflammable aerosol that delivers freezing power exactly where needed. It cools to –65 °F (18.3 °C), dries fast, and leaves no residue. This freeze spray is ideal for lowering the temperature on electrical components to test intermittent faults, detect defective transistors, and provide protection during soldering. Saves time and eliminates water crystals caused by slower freezing methods.

Caution: Contact with skin can cause irritation or frostbite.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR