XFD488 NHS Ester fluorescent probe
Supplier: AAT BIOQUEST INC
|
Warning
|
71902
1812
77463-078EA
1303.8
CAD
76484-514
77463-078
XFD488 NHS Ester fluorescent probe
XFD488 NHS Ester
XFD488 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 488 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Caution: For research use only.
MW:
643.4 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...