XFD488 NHS Ester, Ultra Pure Grade fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

71901 1810
77463-076EA 1649.71 CAD
77463-076 76484-854
XFD488 NHS Ester, Ultra Pure Grade fluorescent probe
XFD488 NHS Ester

XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal
  • Stable for 12 months


Quality tested in-house.


Caution: For research use only.

MW: 643.4 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR