Steri-Wrap® Unite™ SMS Bonded Sterilization Wraps, Propper
Supplier: Propper
31502400
76479-174CS 283.89 CAD
76479-174
Steri-Wrap® Unite™ SMS Bonded Sterilization Wraps, Propper
Sterilization Wraps
Propper Manufacturing Company Steri-Wrap Unite 60 sterilization wraps for simultaneous wrapping of medical devices that will be sterilized via steam, ethylene oxide, and vaporized hydrogen peroxide sterilization.
  • Barrier allows sterilant penetration but prevents penetration of microorganisms
  • Double-wrap protection in half the time compared with sequential double wrapping
  • Strong, tear-resistant 60 gsm material soft enough to enable easy wrapping
  • Dual-color bonded blue/green wrap helps differentiate specific instrument sets
  • Lot code printed on every application
Steri-Wrap Unite 60 sterilization wraps comprise of two sheets of SMS bonded together for convenient simultaneous wrapping of one or a collection of medical devices that will be sterilized following standard healthcare practices. The wraps are made of polypropylene with blue or green pigment and anti-static treatment. Steri-Wrap Unite is used to contain instruments during steam, ethylene oxide, and vaporized hydrogen peroxide sterilization and maintain sterility after processing.
These sterilization wraps allow effective sterilant penetration and maintain the sterility of reusable devices during handling and storage. The two-color sheet allows for easier visualization of damage, tears, or punctures and can be used to differentiate between loaner and facility-owned trays.
Shelf life after sterilization: Steri-Wrap Unite 60 may be used to maintain sterility for 365 days.
Certifications: Steri-Wrap Unite 60 conforms to ISO 11607-1:2019 and EN 868-2:2017.31505400
FDA approved and CE marked